Product Name
Monoclonal Anti-CD203c antibody produced in mouse clone NP4D6, purified immunoglobulin, buffered aqueous solution
Product Number
SAB4700461
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | Report result |
Concentration | Report result |
Purification | Protein A purified |
Purity | > 95% (by SDS-PAGE) |
Western Blotting | Not verified |
Immunofluoresence | Not verified |
Flow Cytometry | The reagent is designed for Flow Cytometry analysis. Suggested working dilution for Flow Cytometry is 1-5 µg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator. |