Product Name
Monoclonal Anti-BECN1 antibody produced in mouse clone 2A4, ascites fluid
Product Number
SAB5300513
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | light yellow liquid |
Concentration | Not Available |
ELISA Antibody Concentration | Antibody concentration: 1/10000, using recombinant protein. |
Western Blotting (Recombinant Protein) | Antibody concentration: 1/500 - 1/2000. |
Western Blotting (Cell Lysate) | Antibody concentration: 1/500 - 1/2000. |
Immunohistochemistry | Antibody concentration: 1/200 - 1/1000. |
Immunofluorescence | Not Determined |
Immunoprecipitation | Not Determined |
Flow Cytometry | Antibody concentration: 1/200 - 1/400. |
Chromatin Immunoprecipitation | Not Determined |