Product Name
Anti-SERCA2/ATP2A2 antibody produced in rabbit
Product Number
SAB5701015
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Concentration | Report result |
Western Blotting | 1:500 - 1:2000 |
Immunoprecepitation | 1:50 - 1:200 |