Product Name
CTGF human recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)
Product Number
SRP4702
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | Lyophilized protein |
SDS-PAGE | =95% |
HPLC | =95% |
Endotoxin | <0.1 ng per mg by LAL method |
Biological Activity | ED50 1.0�2.0 ug/ml determined by the dose-dependent stimulation of the proliferation of HUVEC cells. |