Product Name
Monoclonal Anti-GATA2 antibody produced in mouse clone 2D11, purified immunoglobulin, buffered aqueous solution
Product Number
WH0002624M1
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Western Blotting (Cell lysate) | Antibody concentration: 1 - 5 ug/ml. Specific band of ~50 kDa using Hela NE lysate. |
Immunofluorescence (IF) | Antibody concentration: 10 ug/ml using HeLa cell. |
Sandwich ELISA | Detection limit for recombinant GST tagged GATA2 is approximately 0.03 ng/ml. |
ELISA (Recombinant protein) | Antibody concentration: 1 ug/ml using recombinant protein. |
Western Blotting (Recombinant protein) | Antibody concentration: 1 ug/ml. Specific band of ~37 kDa using immunogen protein lysate. |