Product Name
Monoclonal Anti-TFPI2 antibody produced in mouse clone 3E8, purified immunoglobulin, buffered aqueous solution
Product Number
WH0007980M1
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Immunohistochemistry-Paraffin Embedded Tissue (IHC-P) | Antibody concentration: 1 ug/ml using human tonsil tissue. |
Sandwich ELISA | Detection limit for recombinant GST tagged TFPI2 is approximately 0.03ng/ml. |
ELISA (Recombinant protein) | Antibody concentration: 1 ug/ml using recombinant protein. |