Product Name
Decanoic acid-1,2,3,4-13C4 analytical standard
Product Number
05346
Product Brand
SUPELCO
CAS Number
2561494-72-0
Molecular Weight
176.24
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless and White to Off White |
APPEARANCE (FORM) | Powder or Crystal or Chunk(s) or Solid |
PURITY (GC AREA %) | ≥ 98.0 % |
IDENTIFICATION (LC-MS) | Identity confirmed |
REMARKS ON LC-MS | Degree of Labelling ≥ 99 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
CARBON 13 NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 36 MONTHS |