Skip to Content
Merck
1-Cyclopropyl-7-(4-ethylpiperazin-1-yl)-6-fluoroquinolin-4(1H)-one certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

1-Cyclopropyl-7-(4-ethylpiperazin-1-yl)-6-fluoroquinolin-4(1H)-one

1-Cyclopropyl-7-(4-ethylpiperazin-1-yl)-6-fluoroquinolin-4(1H)-one certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Empirical Formula (Hill Notation):
C18H22FN3O
CAS Number:
Molecular Weight:
315.39

Products

Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Special Grade