
Alternariol-9-methyl ether solution

Empirical Formula (Hill Notation):
C15H12O5
CAS Number:
Molecular Weight:
272.25
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Technique
Showing 1-1 of 1