Skip to Content
Merck
Alternariol solution 20 μg/g in acetonitrile (nominal concentration), certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Alternariol solution

Alternariol solution 20 μg/g in acetonitrile (nominal concentration), certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
3,7,9-Trihydroxy-1-methyl-6H-dibenzo[b,d]pyran-6-one solution
Empirical Formula (Hill Notation):
C14H10O5
CAS Number:
Molecular Weight:
258.23

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Technique