Skip to Content
Merck
AMPK Signaling Activator XV

AMPK Signaling Activator XV

AMPK Signaling Activator XV
Synonym(s):
A-769662, 4-Hydroxy-3-(2′-hydroxy-1,1′-biphenyl-4-yl)-6-oxo-6,7-dihydrothieno[2,3-b]pyridine-5-carbonitrile, 4-Hydroxy-3-(2′-hydroxybiphenyl-4-yl)-6-oxo-6,7-dihydrothieno[2,3-b]pyridine-5-carbonitrile, 6,7-Dihydro-4-hydroxy-3-(2′-hydroxy[1,1′-biphenyl]-4-yl)-6-oxothieno[2,3-b]pyridine-5-carbonitrile, A 769662, A769662
Empirical Formula (Hill Notation):
C20H12N2O3S
CAS Number:
Molecular Weight:
360.39

Products

Brand
Formula Weight
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing