Skip to Content
IN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Safety Data Sheets (SDS)
Certificates (COA/COO)
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
microcentrifuge tubes, 1.5mL with lid, polypropylene
Products
Product Category
centrifuges (6)
tubes (6)
vials (6)
Feature
Manufacturer
Sterilization Compatibility
Working Volume (ml)
Cap
Color
Package Size
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
BR780420
BRAND
®
microcentrifuge tubes, 1.5mL with lid, polypropylene,
transparent, RCF max 20k G, BIO-CERT
®
PCR QUALITY, 500EA
Expand
View Pricing
BR780421
BRAND
®
microcentrifuge tubes, 1.5mL with lid, polypropylene,
yellow, RCF max 20k G, BIO-CERT
®
PCR QUALITY, 500EA
Expand
View Pricing
BR780422
BRAND
®
microcentrifuge tubes, 1.5mL with lid, polypropylene,
blue, RCF max 20k G, BIO-CERT
®
PCR QUALITY, 500EA
Expand
View Pricing
BR780423
BRAND
®
microcentrifuge tubes, 1.5mL with lid, polypropylene,
green, RCF max 20k G, BIO-CERT
®
PCR QUALITY, 500EA
Expand
View Pricing
BR780424
BRAND
®
microcentrifuge tubes, 1.5mL with lid, polypropylene,
orange, RCF max 20k G, BIO-CERT
®
PCR QUALITY, 500EA
Expand
View Pricing
BR780425
BRAND
®
microcentrifuge tubes, 1.5mL with lid, polypropylene,
amber, RCF max 20k G, BIO-CERT
®
PCR QUALITY, 500EA
Expand
View Pricing