Skip to Content
IN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Safety Data Sheets (SDS)
Certificates (COA/COO)
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
Transferpette
®
12-channel, digital adjustable pipette
Products
Product Category
pipettors (7)
Manufacturer
Volume (µL)
Electronic
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
Z328197
BRAND
®
Transferpette
®
12-channel, digital adjustable pipette,
volume 5-50 μL, accuracy: 0.8%, precision: 0.4%
Expand
View Pricing
Z328219
BRAND
®
Transferpette
®
12-channel, digital adjustable pipette,
volume 20-200 μL, accuracy: 0.6%, precision: 0.3%
Expand
View Pricing
Z328162
BRAND
®
Transferpette
®
12-channel, digital adjustable pipette,
volume 0.5-10 μL, accuracy: 1.6%, precision: 1.0%
Expand
View Pricing
Z328227
BRAND
®
Transferpette
®
12-channel, digital adjustable pipette,
volume 30-300 μL, accuracy: 0.6%, precision: 0.3%
Expand
View Pricing
Z328189
BRAND
®
Transferpette
®
12-channel, digital adjustable pipette,
volume 2.5-25 μL, accuracy: 1.0%, precision: 0.6%
Expand
View Pricing
Z328200
BRAND
®
Transferpette
®
12-channel, digital adjustable pipette,
volume 10-100 μL, accuracy: 0.8%, precision: 0.3%
Expand
View Pricing
Z328170
BRAND
®
Transferpette
®
12-channel, digital adjustable pipette,
volume 2-20 μL, accuracy: 1.0%, precision: 0.6%
Expand
View Pricing