Skip to Content
Merck
Buffered NaCl Peptone Solution bottle capacity 250 mL, bottle filling volume 200 mL, closure type, Blue crimp cap with septum, pack of 6 bottles

Buffered NaCl Peptone Solution

Buffered NaCl Peptone Solution bottle capacity 250 mL, bottle filling volume 200 mL, closure type, Blue crimp cap with septum, pack of 6 bottles
Synonym(s):
Sodium Chloride Peptone Buffer

Products

Product Category
Brand
Physical Form
Agency
Capacity (mL)
Quality Segment
Closure Type
Application
Package Size
Sterility
Sterilization Method
Technique
Showing 1-14 of 14
Compare
Product Number
Description
Pricing