
Buffered NaCl-Peptone Solution with LTH

Synonym(s):
Sodium Chloride Peptone Buffer
Products
Product Category
Brand
Physical Form
Agency
Capacity (mL)
Quality Segment
Closure Type
Application
Package Size
Sterility
Sterilization Method
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing