Skip to Content
Merck
vibrant-m

Cabazitaxel acetone CRS

vibrant-m
Synonym(s):
Cabazitaxel acetone
Empirical Formula (Hill Notation):
C45H57NO14·C3H6O
CAS Number:
Molecular Weight:
894.01

Products

Formula Weight
Manufacturer
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing