
Chlorfenapyr-(phenyl-13C6)

Synonym(s):
4-Bromo-2-(4-chlorophenyl-13C6)-1-(ethoxymethyl)-5-(trifluoromethyl)-1H-pyrrol-3-carbonitrile, Chlorfenapyr-13C6
Empirical Formula (Hill Notation):
13C6C9H11BrClF3N2O
Molecular Weight:
413.57
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing