Skip to Content
IN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Safety Data Sheets (SDS)
Certificates (COA/COO)
Quality & Regulatory
Calculators & Apps
Webinars
Back
Greiner FLUOTRAC
™
200 microplate
Products
Product Category
plates (3)
Manufacturer
Wells
Color
Package Size
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
GN655076
Greiner FLUOTRAC
™
200 microplate,
96 well, flat bottom, medium binding, black
Expand
View Pricing
GN781076
Greiner FLUOTRAC
™
200 microplate,
384 well, flat bottom, medium binding, black
Expand
View Pricing
GN784076
Greiner FLUOTRAC
™
200 microplate,
384 well, HiBase, medium binding, Small Volume
™
, black
Expand
View Pricing