Skip to Content
IN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Safety Data Sheets (SDS)
Certificates (COA/COO)
Quality & Regulatory
Calculators & Apps
Webinars
Back
Greiner MASTERBLOCK
®
storage microplate
Products
Product Category
plates (5)
Manufacturer
Wells
Package Size
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
GN780271
Greiner MASTERBLOCK
®
storage microplate,
96 well, polypropylene, capacity 2.0 mL, conical (V) bottom, translucent
Expand
View Pricing
GN780270
Greiner MASTERBLOCK
®
storage microplate,
96 well, polypropylene, capacity 2.0 mL, conical (V) bottom, translucent
Expand
View Pricing
GN786261
Greiner MASTERBLOCK
®
storage microplate,
Deep Well, 96 well, polypropylene, capacity 0.5 mL, conical (V) bottom, clear
Expand
View Pricing
GN780285
Greiner MASTERBLOCK
®
storage microplate,
96 well, polypropylene, capacity 2.0 mL, conical (V) bottom, translucent
Expand
View Pricing
GN781270
Greiner MASTERBLOCK
®
storage microplate,
Deep Well, 384 well, polypropylene, capacity 240 uL, conical (V) bottom, translucent
Expand
View Pricing