
Heptane-1-sulfonic acid sodium salt monohydrate

Synonym(s):
Heptane-1-sulfonic acid sodium salt
Empirical Formula (Hill Notation):
C7H15NaO3S
CAS Number:
Molecular Weight:
202.25
Products
Brand
Formula Weight
Product Line
Showing 1-1 of 1
Compare
Product Number
Description
Pricing