Skip to Content
Merck
Hexane-1-sulphonic Acid Sodium Salt Monohydrate

Hexane-1-sulphonic Acid Sodium Salt Monohydrate

Hexane-1-sulphonic Acid Sodium Salt Monohydrate
Synonym(s):
1-Hexanesulfonic Acid, Sodium Salt, Monohydrate
Linear Formula:
CH3(CH2)5SO3Na · H2O
CAS Number:

Products

Brand
Showing 1-1 of 1
Compare
Product Number
Description
Pricing