Skip to Content
Merck
Lithium bis(oxalate)borate ≥99.9% trace metals basis, ≤100 ppm H2O, battery grade

Lithium bis(oxalate)borate

Lithium bis(oxalate)borate ≥99.9% trace metals basis, ≤100 ppm H2O, battery grade
Synonym(s):
Lithium bis(oxalato)borate, LiBOB, Lithium bis(ethanedioato)borate, Lithium bis(oxalate)borate
Linear Formula:
LiB(C2O4)2
CAS Number:
Molecular Weight:
193.79

Products

Brand
Feature
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Showing 1-1 of 1