
Nitrothal-isopropyl

Synonym(s):
Diisopropyl 5-nitroisophthalate
Empirical Formula (Hill Notation):
C14H17NO6
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2