Skip to Content
Merck
Oxitropium bromide impurity B European Pharmacopoeia (EP) Reference Standard

Oxitropium bromide impurity B

Oxitropium bromide impurity B European Pharmacopoeia (EP) Reference Standard
Synonym(s):
(−)-Scopolamine methyl bromide, Hyoscine methyl bromide, Methscopolamine bromide
Empirical Formula (Hill Notation):
C18H24NO4 · Br
CAS Number:
Molecular Weight:
398.29

Products

Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing