Skip to Content
Merck
vibrant-m

Penbutolol sulfate

vibrant-m
Empirical Formula (Hill Notation):
C36H60N2O8S
CAS Number:
Molecular Weight:
680.94

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing