Product Name
Diethylamine hydrochloride ReagentPlus®, 99%
Product Number
127744
Product Brand
ALDRICH
CAS Number
660-68-4
Molecular Weight
109.60
Test | Specification |
---|---|
Appearance (Color) | White |
Appearance (Form) | Conforms to Requirements |
Powder, Crystals or Crystalline Powder | |
Infrared spectrum | Conforms to Structure |
Titration by AgNO3 | 98.5 - 101.5 % |
GC (area %) | ≥98.5 % |
Solubility (Turbidity) | Clear |
50mg/ml, H2O | |
Solubility (Color) | Colorless |
Recommended Retest Period: 4 Years | Conforms |