Product Name
Bromhexine hydrochloride ≥98.0% (AT)
Product Number
17343
Product Brand
ALDRICH
CAS Number
611-75-6
Molecular Weight
412.59
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless or White |
APPEARANCE (FORM) | Powder |
ARGENTOMETR. TITRATION | 98.0 - 102.0 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |