Product Name
(S)-(+)-2-Phenylbutyric acid 99%
Product Number
279889
Product Brand
ALDRICH
CAS Number
4286-15-1
Molecular Weight
164.20
Test | Specification |
---|---|
APPEARANCE | VISCOUS COLORLESS LIQUID |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 98.5% - 101.5% (WITH NAOH) |
OPTICAL ROTATION | +92(+/- 5) DEGREES (C=0.9%, TOLUENE) |
GAS LIQUID | 98.5% (MINIMUM) |
APPROVED FEBRUARY 8, 2002 JSB |