Product Name
(1S,2S)-1,2-Bis(4-methoxyphenyl)ethylenediamine dihydrochloride 96%
Product Number
684120
Product Brand
ALDRICH
CAS Number
820965-96-6
Molecular Weight
345.26
Test | Specification |
---|---|
Appearance (Color) | White to Tan |
Appearance (Form) | Conforms |
Solid, Powder, Crystals, Crystalline Powder and/or | |
Chunks | |
Infrared spectrum | Conforms to Structure |
Titration by AgNO3 | 95.5 - 104.5 % |
Optical Rotation | 8.0 - 14.0 |
Degrees C=1%, H2O |