Product Name
4-Methylpiperidine 96%
Product Number
M73206
Product Brand
ALDRICH
CAS Number
626-58-4
Molecular Weight
99.17
Test | Specification |
---|---|
APPEARANCE | COLORLESS TO YELLOW LIQUID |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 95.5%-104.5% (WITH HCLO4) |
GAS LIQUID | 95.5% (MINIMUM) |
COLOR TEST | 300 APHA (MAXIMUM) |
APPROVED OCTOBER 11, 2005 RJM |