Product Name
Tri-O-acetyl-D-glucal 98%
Product Number
T44407
Product Brand
ALDRICH
CAS Number
2873-29-2
Molecular Weight
272.25
Test | Specification |
---|---|
Appearance (Color) | White to Off-White |
White to Off-White | |
Appearance (Form) | Conforms to Requirements |
Powder to Powder and/or Chunks | |
Infrared spectrum | Conforms to Structure |
Optical Rotation | -26.0 - -22.0 |
C = 1%, CHCl3 | |
Purity (GC) | ≥97.5 % |
Solubility (Turbidity) | Clear to Very Slightly Hazy |
Solubility (Color) | Colorless to Light Yellow |
5% in CHCl3 |