Product Name
Chlorimuron ethyl PESTANAL®, analytical standard
Product Number
32874
Product Brand
SIAL
CAS Number
90982-32-4
Molecular Weight
414.82
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Light Yellow |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 95.0 % |
SOLUBILITY (COLOR) | Colorless to Light Yellow |
SOLUBILITY (TURBIDITY) | Clear |
SOLUBILITY (METHOD) | 10 MG in 10 ML ACTONITRILE |
WATER | ≤ 1.0 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 60 MONTHS |