Product Name
Triclabendazole sulfoxide VETRANAL®, analytical standard
Product Number
35391
Product Brand
SIAL
CAS Number
100648-13-3
Molecular Weight
375.66
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Off White |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 99.0 % |
MELTING POINT | REPORT RESULT |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
RESIDUAL SOLVENTS | ≤ 1.5 % |
EXPIRY PERIOD | 60 MONTHS |