Product Name
Sodium perchlorate monohydrate suitable for HPLC, LiChropur™, 99.0-101.0% (T)
Product Number
56581
Product Brand
SIAL
CAS Number
7791-07-3
Molecular Weight
140.46
Test | Specification |
---|---|
APPEARANCE (COLOR) | White |
APPEARANCE (FORM) | Powder or Crystals |
TITRATION (ION EXCHANGE) | 99.0 - 101.0 % |
SOLUBILITY (COLOR) | Colorless |
SOLUBILITY (TURBIDITY) | Clear |
SOLUBILITY (METHOD) | 5% IN WATER |
PH | 6.0 - 8.0 |
UV Absorbance | UV-Absorption at 210 nm ≤ 0.100 |
UV Absorbance | UV-Absorption at 220 nm ≤ 0.050 |
UV Absorbance | UV-Absorption at 230 nm ≤ 0.030 |
UV Absorbance | UV-Absorption at 240 nm ≤ 0.020 |
UV Absorbance | UV-Absorption at 500 nm ≤ 0.020 |
CYCLIC VOLTAMMOGRAM | CORRESPONDS TO STANDARD CV |
RESIDUE (FILTER TEST) | NO RESIDUE |
FILTERTEST (METHOD) | 10% IN WATER |
RECOMMENDED RETEST PERIOD | 54 MONTHS |