Product Name
Anti-MSH3 (center) antibody produced in rabbit saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
Product Number
SAB1300711
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | colorless liquid |
Concentration | >= 0.25 mg/ml |
ELISA | 1:1000 |
Western Blotting | 1:100-1:500 |
Immunohistochemistry | Not Verified |
Immunofluorescence | Not Verified |
Immunoprecipitation | Not Verified |
Flow Cytometry | Not Verified |
Chromatin Immunoprecipitation | Not Verified |