Product Name
Anti-F3 antibody produced in rabbit purified immunoglobulin, buffered aqueous solution
Product Number
SAB1410355
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | Report result |
Concentration | Report result |
Western Blot(Tissue lysate) | Antibody concentration: 1 ~ 5 µg/mL. Specific band of ~33.1 kDa using human kidney lysate |
Western Blotting (Transfected lysate) | Antibody concentration: 0.1 ~ 5 µg/mL. Specific band of ~33.1 kDa using F3 transfected 293T lysate |