Product Name
Anti-Chicken IgG (H+L)-Alkaline Phosphatase antibody produced in goat affinity isolated antibody, buffered aqueous solution
Product Number
SAB3700201
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | Report result |
Concentration | Report result |
Purification | Affinity Purified |
ELISA | 1:2,000 - 1:10,000 |
Western Blotting | 1:500 - 1:2,500 |
Immunohistochemistry | 1:200 - 1:1,000 |