Product Name
Monoclonal Anti-CD22-PE antibody produced in mouse clone MEM-01, purified immunoglobulin, buffered aqueous solution
Product Number
SAB4700130
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | Report result |
Concentration | Content of a vial (2 mL) is sufficient for 100 tests. |
Purification | The purified antibody is conjugated with R-Phycoerythrin (PE) under optimum conditions. The conjugate is purified by size-exclusion chromatography and adjusted for direct use. No reconstitution is necessary. |
Purity | N/A |
Western Blotting | Not verified |
Immunofluoresence | Not verified |
Flow Cytometry | The reagent is designed for Flow Cytometry analysis of human blood cells using 20 µL reagent / 100 µL of whole blood or 1e6 cells in a suspension. The content of a vial (2 mL) is sufficient for 100 tests. |