Product Name
Valeryl-L-carnitine-(N-methyl-d3) analytical standard
Product Number
08459
Product Brand
SUPELCO
CAS Number
Molecular Weight
248.33
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Off White |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 95.0 % |
IDENTIFICATION (LC-MS) | Identity Confirmed |
REMARKS ON LC-MS | Degree of Labelling ≥ 99 % |
SPECIFIC ROTATION (20/D) | -22.0 ± 2.0 Degrees |
CONCENTRATION | c=0.1 in water |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 36 MONTHS |