Skip to Content
Merck
1,1,2,2-Tetrafluoro-2-(1,1,2,2,2-pentafluoroethoxy)ethanesulfonic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

1,1,2,2-Tetrafluoro-2-(1,1,2,2,2-pentafluoroethoxy)ethanesulfonic acid

1,1,2,2-Tetrafluoro-2-(1,1,2,2,2-pentafluoroethoxy)ethanesulfonic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Perfluoro-(2-ethoxyethane)sulfonic acid
Empirical Formula (Hill Notation):
C4HF9O4S
CAS Number:
Molecular Weight:
316.10

Products

Brand
Formula Weight
Manufacturer
Quality Segment
Application
Reference Material Type
Special Grade