
1-Caffeoylquinic acid

Synonym(s):
1-O-Caffeoylquinic acid, 1-O-(3,4-Dihydroxycinnamoyl)-D-quinic acid
Empirical Formula (Hill Notation):
C16H18O9
CAS Number:
Molecular Weight:
354.31
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing