Skip to Content
Merck
vibrant-m

5-[2-Ethoxy-5-[(4-methyl-4-oxido-1-piperazinyl)sulfonyl]phenyl]-1,6-dihydro-1-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidin-7-one

vibrant-m
Synonym(s):
1-Methyl-4-[[4-ethoxy-3-(1-methyl-7-oxo-3-propyl-6,7-dihydro-1H-pyrazolo[4,3-d]pyrimidin-5-yl)phenyl]sulfonyl]piperazine-1-oxide, Sildenafil-N-oxide
Empirical Formula (Hill Notation):
C22H30N6O5S
CAS Number:
Molecular Weight:
490.58

Products

Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Special Grade