
Germacrone

Synonym(s):
(E,E)-Germacra-3,7(11),9-trien-6-one, trans,trans-3,7-Dimethyl-10-isopropylidene-3,7-cyclodecadien-1-one
Empirical Formula (Hill Notation):
C15H22O
CAS Number:
Molecular Weight:
218.33
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing