Skip to Content
Merck
Lercanidipine hydrochloride ≥98% (HPLC)

Lercanidipine hydrochloride

Lercanidipine hydrochloride ≥98% (HPLC)
Synonym(s):
1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-2-[(3,3-diphenylpropyl)methylamino]-1,1-dimethylethyl methyl ester 3,5-pyridinedicarboxylic acid hydrochloride
Empirical Formula (Hill Notation):
C36H41N3O6 · HCl
CAS Number:
Molecular Weight:
648.19

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1