Skip to Content
Merck
Monoclonal Anti-Rat Kappa Light Chains (1a + 1b) antibody produced in mouse clone RT-39, ascites fluid

Monoclonal Anti-Rat Kappa Light Chains (1a + 1b) antibody produced in mouse

Monoclonal Anti-Rat Kappa Light Chains (1a + 1b) antibody produced in mouse clone RT-39, ascites fluid
Synonym(s):
Monoclonal Anti-Rat Kappa Light Chains (1a + 1b)

Products

Product Category
Brand
Biological Source
Clonality
Isotype
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Technique
Showing 1-1 of 1