Skip to Content
Merck
Nonylphenol monoethoxylate solution 50 μg/mL in acetone, technical, analytical standard

Nonylphenol monoethoxylate solution

Nonylphenol monoethoxylate solution 50 μg/mL in acetone, technical, analytical standard
Synonym(s):
NP-1EO technical
Empirical Formula (Hill Notation):
C17H28O2
Molecular Weight:
264.40
EC Number:

Products

Brand
Formula Weight
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1