Skip to Content
Merck
vibrant-m

Potassium 2-[(6-chloro-1,1,2,2,3,3,4,4,5,5,6,6-dodecafluorohexyl)oxy]-1,1,2,2-tetrafluoroethanesulfonate

vibrant-m
Synonym(s):
6:2 Cl-PFAES
Empirical Formula (Hill Notation):
C8ClF16KO4S
CAS Number:
Molecular Weight:
570.67

Products

Brand
Feature
Formula Weight
Manufacturer
Quality Segment
Reference Material Type
Special Grade