Skip to Content
Merck
Sucralose granular EMPROVE® ESSENTIAL Ph Eur,ChP,NF,JPE | 56038-13-2 granular, EMPROVE® ESSENTIAL, Ph. Eur., JPE, NF

Sucralose granular EMPROVE® ESSENTIAL Ph Eur,ChP,NF,JPE | 56038-13-2

Sucralose granular EMPROVE® ESSENTIAL Ph Eur,ChP,NF,JPE | 56038-13-2 granular, EMPROVE® ESSENTIAL, Ph. Eur., JPE, NF
Synonym(s):
Sucralose, 1,6-Dichloro-1,6-dideoxy-β-D-fructofuranosyl-4-chloro-4-deoxy-α-D-galactopyranoside, E955, Trichlorosucrose
Empirical Formula (Hill Notation):
C12H19Cl3O8
CAS Number:
Molecular Weight:
397.63

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Application
Particle Size (µm)