
Tetradecamethylhexasiloxane

Empirical Formula (Hill Notation):
C14H42O5Si6
CAS Number:
Molecular Weight:
458.99
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing