Merck
  • Ischemia-modified albumin levels in patients with end-stage renal disease patients on hemodialysis: does albumin analysis method affect albumin-adjusted ischemia-modified albumin levels?
Ischemia-modified albumin levels in patients with end-stage renal disease patients on hemodialysis: does albumin analysis method affect albumin-adjusted ischemia-modified albumin levels?
Journal of clinical laboratory analysis (2010-07-14)
Aysel Kiyici, Idris Mehmetoğlu, Hatice Karaoğlan, Hüseyin Atalay, Yalçin Solak, Süleyman Türk
ABSTRACT
Ischemia-Modified albumin (IMA) has been used as an early marker in the evaluation of the patients with acute coronary syndrome. We aimed to evaluate IMA in end-stage renal disease (ESRD) patients on hemodialysis and the effect of albumin methods on albumin-adjusted IMA levels. A total of 30 ESRD patients were included in this study. Serum IMA and albumin levels were measured before and after a hemodialysis session. Albumin concentrations were determined with bromocresol green and bromocresol purple methods. Postdialysis IMA and albumin-adjusted IMA levels with two different albumin methods were significantly increased compared with the predialysis levels (P<0.05). However, we did not find any difference in albumin-adjusted IMA levels in either at the beginning or at the end of the dialysis session. IMA levels increase after hemodialysis, whereas albumin method has no effect on albumin-adjusted IMA levels.
Sigma-Aldrich
Bromocresol Green Sultone Form
Sigma-Aldrich
Bromocresol Green sodium salt, 90% (HPLC), ACS reagent, Dye content 90 %
Sigma-Aldrich
Bromocresol Purple, Technical grade
Sigma-Aldrich
Bromocresol Purple, BioReagent, suitable for indicator, Dye content 90 %
Sigma-Aldrich
Bromocresol Green, ACS reagent, Dye content 95 %